Assay Details
| Gene Symbol: Cox8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007751.3 | NP_031777.1 | 1 - 2 | 213 | 79 | |
| GenBank mRNA | AA028501.1 | - | 1 - 2 | 190 | 79 | |
| AA511205.1 | - | 1 - 2 | 167 | 79 | ||
| AK003116.1 | - | 1 - 2 | 156 | 79 | ||
| BC027531.1 | - | 1 - 2 | 157 | 1515187 | 79 | |
| BC086930.1 | - | 1 - 2 | 176 | 6752860 | 79 | |
| BE626379.1 | - | 1 - 2 | 213 | 79 | ||
| U15541.1 | - | 1 - 2 | 163 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P423814 | NM_007751.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P423814 | AA028501.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P423814 | AA511205.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P423814 | AK003116.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P423814 | BC027531.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P423814 | NM_007751.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P423814 | AA028501.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P423814 | AA511205.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P423814 | AK003116.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P423814 | BC027531.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 628590 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 941232 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 124685 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P173008 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P423814 | NM_007751.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P423814 | AA028501.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P423814 | AA511205.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P423814 | AK003116.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P423814 | BC027531.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P423814 | NM_007751.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P423814 | AA028501.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P423814 | AA511205.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P423814 | AK003116.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P423814 | BC027531.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P423814 | NM_007751.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P423814 | AA028501.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P423814 | AA511205.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P423814 | AK003116.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P423814 | BC027531.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P423814 | NM_007751.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P423814 | AA028501.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P423814 | AA511205.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P423814 | AK003116.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P423814 | BC027531.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585565 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585571 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585581 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585580 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585558 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585584 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585554 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585578 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585552 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585562 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585551 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585591 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585590 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585572 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585595 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585575 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585587 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585546 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585564 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585563 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585555 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585549 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585550 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585573 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585588 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585560 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585593 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585556 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585559 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585576 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585585 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585569 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585567 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P173008 | - | - |
| Whole Mouse Genome G4122A | A_52_P423814 | NM_007751.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P423814 | AA028501.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P423814 | AA511205.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P423814 | AK003116.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P423814 | BC027531.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P423814 | NM_007751.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P423814 | AA028501.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P423814 | AA511205.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P423814 | AK003116.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P423814 | BC027531.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133345 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103330 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P173008 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06533537 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06533540 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06533505 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06533554 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06533523 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06533521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585571 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585573 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585582 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585580 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585610 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585564 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585618 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05934704 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585552 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06533512 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585594 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585603 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585560 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585565 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05934703 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585587 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585549 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585555 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585556 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05982772 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06533561 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585562 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585551 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06533501 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585558 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06102503 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585588 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585584 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06243434 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585575 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585585 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585612 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06533514 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585571 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585554 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585572 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585604 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585547 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585599 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585590 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06165226 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06469813 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05982767 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585593 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585608 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10569009 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10569010 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449218_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10569008 | - | - |
| Mouse Genome 430 2.0 Array | 1449218_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1449218_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12869_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12869_PM_at | - | - |
| Affymetrix Murine Genome 6500A Array | Msa17160_i_at | - | - |
| Affymetrix Murine Genome 6500A Array | Msa17160_f_at | - | - |
| Affymetrix Murine Genome 6500A Array | Msa17160_r_i_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700037886.mm.1 | NM_007751.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700037886.mm.1 | NM_007751.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700037886.mm.1 | TC0700004510.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700037887.mm.1 | NM_007751.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700037887.mm.1 | NM_007751.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700037887.mm.1 | TC0700004510.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700037889.mm.1 | NM_007751.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700037889.mm.1 | NM_007751.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700037889.mm.1 | TC0700004510.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1449218_at | - | - |
| Mouse Expression Set 430 | 1449218_PM_at | - | - |
| Murine Genome U74v2 Set | 161205_at | - | - |
| Murine Genome U74v2 Set | 160851_PM_r_at | - | - |
| Murine Genome U74v2 Set | 160851_r_at | - | - |
| Murine Genome U74v2 Set | 161205_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449218_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449218_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449218_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449218_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 608352 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2360280 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002360280 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2727520 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2727520 | - | - |
| mouse-6 v1.1 (Illumina) | scl30508.2.1_18-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2360280 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl30508.2.1_18-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2360280 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2727520 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2727520 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl30508.2.1_18-S | - | - |

Cox8b
Genomic Map