Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700013404.mm.1 |
NM_007865.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013404.mm.1 |
NM_007865.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013404.mm.1 |
TC1700001497.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700013407.mm.1 |
NM_007865.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013407.mm.1 |
NM_007865.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013407.mm.1 |
TC1700001497.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700013408.mm.1 |
NM_007865.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013408.mm.1 |
NM_007865.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013408.mm.1 |
TC1700001497.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700013410.mm.1 |
NM_007865.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013410.mm.1 |
NM_007865.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013410.mm.1 |
TC1700001497.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700013409.mm.1 |
NM_007865.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013409.mm.1 |
NM_007865.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013409.mm.1 |
TC1700001497.mm.1 |
MTA TC |