Assay Details
Gene Symbol: Dll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007865.3 | NP_031891.2 | 8 - 9 | 1829 | 76 | |
GenBank mRNA | BC057400.1 | - | 8 - 9 | 1342 | 6402691 | 76 |
BC065063.1 | - | 8 - 9 | 1593 | 6416191 | 76 | |
X80903.1 | - | 8 - 9 | 1259 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
