Assay Details
| Gene Symbol: E2f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007891.5 | NP_031917.1 | 1 - 2 | 405 | 80 | |
| GenBank mRNA | AF483514.1 | - | 1 - 2 | 247 | 80 | |
| AF483515.1 | - | 1 - 2 | 247 | 80 | ||
| AK153968.1 | - | 1 - 2 | 370 | 80 | ||
| BC052160.1 | - | 1 - 2 | 347 | 6330425 | 80 | |
| L21973.1 | - | 1 - 2 | 365 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

E2f1
Genomic Map