Assay Details
Gene Symbol: Epha4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007936.3 | NP_031962.2 | 12 - 13 | 2193 | 111 | |
GenBank mRNA | AK028509.1 | - | 12 - 13 | 2186 | 111 | |
AK147698.1 | - | 12 - 13 | 2189 | 111 | ||
BC004782.1 | - | 12 - 13 | 2108 | 3588285 | 111 | |
BC052164.1 | - | 12 - 13 | 2165 | 6512978 | 111 | |
S57168.1 | - | 12 - 13 | 2193 | 111 | ||
X65138.1 | - | 12 - 13 | 2193 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
