Assay Details
| Gene Symbol: Pdia3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007952.2 | NP_031978.2 | 10 - 11 | 1408 | 87 | |
| GenBank mRNA | AK088721.1 | - | 10 - 11 | 1368 | 87 | |
| AK132459.1 | - | 10 - 11 | 1373 | 87 | ||
| AK133799.1 | - | 10 - 11 | 1367 | 87 | ||
| AK167807.1 | - | 10 - 11 | 1361 | 87 | ||
| AK169611.1 | - | 10 - 11 | 1362 | 87 | ||
| BC003285.1 | - | 10 - 11 | 1327 | 3582199 | 87 | |
| BC033439.1 | - | 10 - 11 | 1302 | 4016310 | 87 | |
| M73329.1 | - | 10 - 11 | 1347 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pdia3
Genomic Map