Assay Details
| Gene Symbol: Esr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB560752.1 | - | 1 - 2 | 136 | 102 | |
| AK054182.1 | - | 1 - 2 | 140 | 102 | ||
| AK087638.1 | - | 1 - 2 | 88 | 102 | ||
| M38651.1 | - | 1 - 2 | 118 | 102 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB560752.1, M38651.1 transcripts.
Set Membership: |
|

Esr1
Genomic Map