Assay Details
| Gene Symbol: Esr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB560752.1 | - | 3 - 4 | 857 | 67 | |
| AB560753.1 | - | 2 - 3 | 720 | 67 | ||
| AB560754.1 | - | 2 - 3 | 720 | 67 | ||
| AB560755.1 | - | 2 - 3 | 720 | 67 | ||
| AB560756.1 | - | 2 - 3 | 720 | 67 | ||
| AK036627.1 | - | 3 - 4 | 831 | 67 | ||
| AK039911.1 | - | 6 - 7 | 1334 | 67 | ||
| AK041525.1 | - | 4 - 5 | 979 | 67 | ||
| AK054182.1 | - | 3 - 4 | 862 | 67 | ||
| AK077236.1 | - | 3 - 4 | 836 | 67 | ||
| AK139745.1 | - | 4 - 5 | 1391 | 67 | ||
| EU791538.1 | - | 3 - 4 | 775 | 67 | ||
| EU791540.1 | - | 3 - 4 | 900 | 67 | ||
| M38651.1 | - | 3 - 4 | 839 | 67 | ||
More Information
Set Membership: |
|

Esr1
Genomic Map