Assay Details
| Gene Symbol: Faf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007983.2 | NP_032009.2 | 15 - 16 | 1892 | 79 | |
| GenBank mRNA | AK053752.1 | - | 15 - 16 | 1827 | 79 | |
| BC065098.1 | - | 15 - 16 | 1892 | 30362116 | 79 | |
| U39643.1 | - | 15 - 16 | 1902 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Faf1
Genomic Map