Assay Details
| Gene Symbol: Fgd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008001.4 | NP_032027.2 | 13 - 14 | 2555 | 72 | |
| GenBank mRNA | AK148151.1 | - | 13 - 14 | 2556 | 72 | |
| AK190384.1 | - | 3 - 4 | 148 | 72 | ||
| BC004838.1 | - | 5 - 6 | 445 | 72 | ||
| BC011462.1 | - | 13 - 14 | 2492 | 3674293 | 72 | |
| U22325.1 | - | 13 - 14 | 2154 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fgd1
Genomic Map