Assay Details
| Gene Symbol: Gcgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008101.2 | NP_032127.2 | 4 - 5 | 460 | 62 | |
| GenBank mRNA | AK144393.1 | - | 4 - 5 | 407 | 62 | |
| BC031885.1 | - | 4 - 5 | 410 | 5137340 | 62 | |
| BC057988.1 | - | 4 - 5 | 391 | 5100574 | 62 | |
| L38613.1 | - | 3 - 4 | 62 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Gcgr
Genomic Map