Assay Details
Gene Symbol: Gcgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008101.2 | NP_032127.2 | 6 - 7 | 684 | 69 | |
GenBank mRNA | AK004890.1 | - | 4 - 5 | 810 | 69 | |
AK144393.1 | - | 6 - 7 | 631 | 69 | ||
BC012718.1 | - | 5 - 6 | 759 | 69 | ||
BC031885.1 | - | 6 - 7 | 634 | 5137340 | 69 | |
BC057988.1 | - | 6 - 7 | 615 | 5100574 | 69 | |
L38613.1 | - | 5 - 6 | 594 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
