Assay Details
| Gene Symbol: Gcgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008101.2 | NP_032127.2 | 8 - 9 | 1004 | 103 | |
| GenBank mRNA | AK004890.1 | - | 5 - 6 | 1211 | 103 | |
| AK144393.1 | - | 8 - 9 | 951 | 103 | ||
| BC012718.1 | - | 7 - 8 | 1079 | 103 | ||
| BC031885.1 | - | 8 - 9 | 954 | 5137340 | 103 | |
| BC057988.1 | - | 8 - 9 | 935 | 5100574 | 103 | |
| L38613.1 | - | 7 - 8 | 914 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gcgr
Genomic Map