Assay Details
| Gene Symbol: Gcgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008101.2 | NP_032127.2 | 9 - 10 | 1062 | 66 | |
| GenBank mRNA | AK004890.1 | - | 6 - 7 | 1269 | 66 | |
| AK144393.1 | - | 9 - 10 | 1009 | 66 | ||
| BC012718.1 | - | 8 - 9 | 1137 | 66 | ||
| BC031885.1 | - | 9 - 10 | 1012 | 5137340 | 66 | |
| BC057988.1 | - | 9 - 10 | 993 | 5100574 | 66 | |
| L38613.1 | - | 8 - 9 | 972 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gcgr
Genomic Map