Assay Details
Gene Symbol: Gif | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008118.3 | NP_032144.2 | 3 - 4 | 464 | 93 | |
GenBank mRNA | AK078933.1 | - | 3 - 4 | 465 | 93 | |
BC118519.1 | - | 3 - 4 | 450 | 40105765 | 93 | |
L24191.1 | - | 3 - 4 | 376 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
