Assay Details
Gene Symbol: Gif | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008118.3 | NP_032144.2 | 5 - 6 | 780 | 72 | |
GenBank mRNA | AK078933.1 | - | 5 - 6 | 781 | 72 | |
BC118519.1 | - | 5 - 6 | 766 | 40105765 | 72 | |
L24191.1 | - | 5 - 6 | 692 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1968488 | NM_008118.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1968488 | AK078933.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 637136 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 22178 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 422912 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 732348 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 456221 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 364014 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 100152 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 970587 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 878305 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 50621 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 65502 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164713 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139175 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1968488 | NM_008118.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1968488 | AK078933.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P280697 | NM_008118.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P280697 | AK078933.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P280697 | BC118519.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P280697 | L24191.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957541 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957518 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957522 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957538 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957550 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957544 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957523 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957539 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957546 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957535 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957526 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957532 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957521 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957542 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957551 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957520 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957543 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957536 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31957529 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09107084 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P139175 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P164713 | - | - |
Whole Mouse Genome G4122A | A_51_P280697 | NM_008118.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P280697 | AK078933.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P280697 | BC118519.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P280697 | L24191.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126011 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103849 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P164713 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P139175 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14177178 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957578 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957532 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957680 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957602 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957686 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957691 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957629 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957536 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957622 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957609 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957618 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957565 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957504 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957562 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957696 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957524 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957591 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957574 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957576 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957512 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957514 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957515 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957607 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957643 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957594 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957586 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957645 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957568 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957550 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957692 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957666 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957623 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957698 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957542 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957664 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957552 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957580 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957589 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957570 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957582 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957613 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957632 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957538 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957641 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957679 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957596 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957673 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957668 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957604 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957637 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957560 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957671 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957556 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957584 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957705 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957633 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957704 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957649 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957651 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957626 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957695 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957520 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957644 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957647 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957529 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957681 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957577 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957597 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957554 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957526 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957513 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957653 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957636 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957690 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957566 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957688 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957611 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957539 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957674 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957517 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957544 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957561 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957682 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957600 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957535 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957617 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957541 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957612 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957615 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957634 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957505 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31957610 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957546 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957543 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14294009 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957542 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957536 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957535 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957574 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957539 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957522 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957556 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957514 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14351363 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957558 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957544 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957529 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957517 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957634 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14300136 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957527 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957526 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957550 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957530 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957515 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14451693 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957554 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957563 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957568 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957562 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957572 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957501 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957561 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14184661 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957512 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957557 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14184656 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957538 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957565 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957633 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957552 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957570 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957532 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957632 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957560 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957566 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31957541 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10461661 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10461657 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10461662 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10461658 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10461656 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10461660 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10461659 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10461654 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10461653 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10461655 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1419020_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10461652 | - | - |
Mouse Genome 430 2.0 Array | 1419020_at | - | - |
Mouse Genome 430 2.0 Array | 1419020_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14603_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14603_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1900002810.mm.1 | NM_008118.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900002810.mm.1 | NM_008118.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900002810.mm.1 | TC1900000231.mm.1 | MTA TC |
Mouse Expression Set 430 | 1419020_at | - | - |
Mouse Expression Set 430 | 1419020_PM_at | - | - |
Murine Genome U74v2 Set | 92690_PM_at | - | - |
Murine Genome U74v2 Set | 92690_at | - | - |
Mouse Genome 430A 2.0 Array | 1419020_at | - | - |
Mouse Genome 430A 2.0 Array | 1419020_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419020_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419020_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 402802 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 2260056 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002260056 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2631363 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2631363 | - | - |
mouse-6 v1.1 (Illumina) | scl53354.9.1_26-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2260056 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl53354.9.1_26-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2260056 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2631363 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2631363 | - | - |
