Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100035912.mm.1 |
NM_008122.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035912.mm.1 |
NM_008122.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035912.mm.1 |
TC1100003854.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100035911.mm.1 |
NM_008122.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035911.mm.1 |
NM_008122.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035911.mm.1 |
TC1100003854.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100035908.mm.1 |
NM_001159382.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035908.mm.1 |
NM_001159382.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035908.mm.1 |
NM_001159383.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035908.mm.1 |
NM_001159383.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035908.mm.1 |
NM_008122.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035908.mm.1 |
NM_008122.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035908.mm.1 |
TC1100003854.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100035910.mm.1 |
NM_008122.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035910.mm.1 |
NM_008122.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035910.mm.1 |
TC1100003854.mm.1 |
MTA TC |