Assay Details
| Gene Symbol: Gjb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008126.2 | NP_032152.1 | 1 - 2 | 53 | 87 | |
| GenBank mRNA | X63099.1 | - | 1 - 2 | 89 | 86 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_008126.2,X63099.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23284715 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23284717 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23284716 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23284713 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23284714 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2014124 | BC024387.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2014124 | X63099.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2014124 | NM_001160012.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2014124 | NM_008126.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2014124 | AK083716.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 753763 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 565764 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 736222 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 620394 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 113164 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P128298 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P157477 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P175247 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2014124 | BC024387.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2014124 | X63099.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2014124 | NM_001160012.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2014124 | NM_008126.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2014124 | AK083716.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P353776 | BC024387.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P353776 | X63099.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P353776 | NM_001160012.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P353776 | NM_008126.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P353776 | AK083716.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284768 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284763 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284775 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284778 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284749 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284766 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284767 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284726 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284750 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284738 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284765 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284732 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284740 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284751 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284761 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284743 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284729 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284739 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284773 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284733 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284736 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284727 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284759 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284779 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284748 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284745 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284753 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284752 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284770 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284777 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284756 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284757 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284735 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P157477 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P175247 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P128298 | - | - |
| Whole Mouse Genome G4122A | A_51_P353776 | BC024387.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P353776 | X63099.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P353776 | NM_001160012.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P353776 | NM_008126.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P353776 | AK083716.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113218 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P175247 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P128298 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P157477 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03990368 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03501388 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03501387 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03990367 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03568829 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03568821 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03501385 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03568824 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03990364 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03568830 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03568831 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03501379 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23284713 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23284715 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23284714 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23284717 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23284716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284718 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284783 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03827183 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03326501 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284733 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284736 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284696 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03534487 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03682937 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284699 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284740 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03990368 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284735 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284745 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03973265 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284729 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284704 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284718 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284749 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284782 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03751761 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284701 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03682939 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284763 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284779 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03299923 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284766 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03301101 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284770 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284730 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03568831 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284773 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284715 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284754 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284752 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284716 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284722 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284703 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284756 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284748 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03751754 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284768 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284785 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284706 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284746 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284775 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284751 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284719 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284713 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03501379 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03299913 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284732 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284757 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284780 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284761 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03299925 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10516486 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516485 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416715_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10516484 | - | - |
| Mouse Genome 430 2.0 Array | 1416715_at | - | - |
| Mouse Genome 430 2.0 Array | 1416715_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14620_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14620_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400029033.mm.1 | TC0400003566.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400029033.mm.1 | NM_008126.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400029033.mm.1 | NM_008126.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400029032.mm.1 | TC0400003566.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400029032.mm.1 | NM_001160012.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400029032.mm.1 | NM_001160012.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400029032.mm.1 | NM_008126.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400029032.mm.1 | NM_008126.2 | Refseq |
| Mouse Expression Set 430 | 1416715_at | - | - |
| Mouse Expression Set 430 | 1416715_PM_at | - | - |
| Murine Genome U74v2 Set | 104232_PM_at | - | - |
| Murine Genome U74v2 Set | 104232_at | - | - |
| Murine Genome U74v2 Set | 169540_f_at | - | - |
| Murine Genome U74v2 Set | 169540_PM_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416715_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416715_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416715_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416715_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 923673 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2650040 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2650040 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002650040 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1218561 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1218561 | - | - |
| mouse-6 v1.1 (Illumina) | scl23803.3_247-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2650040 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl23803.3_247-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2650040 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2879102 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2879102 | - | - |

Gjb3
Genomic Map