Assay Details
| Gene Symbol: Gpld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008156.2 | NP_032182.2 | 14 - 15 | 1459 | 69 | |
| GenBank mRNA | AF050666.1 | - | 14 - 15 | 1404 | 69 | |
| AK004957.2 | - | 13 - 14 | 1163 | 69 | ||
| AY081194.1 | - | 13 - 14 | 1186 | 69 | ||
| AY333960.1 | - | 13 - 14 | 1143 | 69 | ||
| BC019146.1 | - | 13 - 14 | 1172 | 5052822 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpld1
Genomic Map