Assay Details
| Gene Symbol: Grin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177656.2 | NP_001171127.1 | 6 - 7 | 1162 | 72 | |
| NM_001177657.2 | NP_001171128.1 | 5 - 6 | 1099 | 72 | ||
| NM_008169.3 | NP_032195.1 | 5 - 6 | 1099 | 72 | ||
| GenBank mRNA | AK158140.1 | - | 5 - 6 | 1039 | 72 | |
| AK158175.1 | - | 6 - 7 | 1100 | 72 | ||
| BC039157.1 | - | 5 - 6 | 1015 | 4507986 | 72 | |
| D10028.1 | - | 5 - 6 | 891 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grin1
Genomic Map