Assay Details
| Gene Symbol: Grin2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008172.2 | NP_032198.2 | 9 - 10 | 2355 | 63 | |
| GenBank mRNA | AB154244.1 | - | 9 - 10 | 2241 | 63 | |
| AB154245.1 | - | 9 - 10 | 2241 | 63 | ||
| D12822.1 | - | 9 - 10 | 2241 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grin2d
Genomic Map