Assay Details
| Gene Symbol: Cfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142706.1 | NP_001136178.1 | 13 - 14 | 2173 | 93 | |
| NM_008198.2 | NP_032224.2 | 13 - 14 | 2173 | 93 | ||
| GenBank mRNA | AK098094.1 | - | 13 - 14 | 1833 | 93 | |
| AK149531.1 | - | 13 - 14 | 1806 | 93 | ||
| AK149573.1 | - | 13 - 14 | 1832 | 93 | ||
| BC005451.1 | - | 13 - 14 | 1807 | 3591669 | 93 | |
| K01496.1 | - | 8 - 9 | 913 | 93 | ||
| M57890.1 | - | 13 - 14 | 1876 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cfb
Genomic Map