Assay Details
| Gene Symbol: Hnf4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008261.2 | NP_032287.2 | 1 - 2 | 229 | 82 | |
| GenBank mRNA | AK143948.1 | - | 1 - 2 | 254 | 82 | |
| BC039220.1 | - | 1 - 2 | 229 | 4238842 | 82 | |
| D29015.1 | - | 1 - 2 | 156 | 82 | ||
| EF193393.1 | - | 1 - 2 | 54 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hnf4a
Genomic Map