Assay Details
| Gene Symbol: Il10ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008348.2 | NP_032374.1 | 1 - 2 | 143 | 73 | |
| GenBank mRNA | AK137663.1 | - | 1 - 2 | 144 | 73 | |
| AK150022.1 | - | 1 - 2 | 140 | 73 | ||
| AK150524.1 | - | 1 - 2 | 147 | 73 | ||
| AK153120.1 | - | 1 - 2 | 147 | 73 | ||
| AK159566.1 | - | 1 - 2 | 140 | 73 | ||
| L12120.1 | - | 1 - 2 | 148 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il10ra
Genomic Map