Assay Details
| Gene Symbol: Il12rb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008353.2 | NP_032379.2 | 13 - 14 | 1789 | 60 | |
| GenBank mRNA | AK137577.1 | - | 13 - 14 | 1790 | 60 | |
| AK171937.1 | - | 13 - 14 | 1763 | 60 | ||
| U23922.1 | - | 13 - 14 | 1744 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il12rb1
Genomic Map