Assay Details
| Gene Symbol: Il12rb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008354.3 | NP_032380.1 | 1 - 2 | 105 | 78 | |
| GenBank mRNA | U64199.1 | - | 1 - 2 | 106 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_008354.3, U64199.1 transcripts.
Set Membership: |
|

Il12rb2
Genomic Map