Assay Details
| Gene Symbol: Il12rb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008354.3 | NP_032380.1 | 7 - 8 | 1145 | 91 | |
| GenBank mRNA | BC137745.1 | - | 6 - 7 | 1057 | 8860764 | 91 |
| U64199.1 | - | 7 - 8 | 1146 | 91 | ||
More Information
Set Membership: |
|

Il12rb2
Genomic Map