Assay Details
| Gene Symbol: Il18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008360.1 | NP_032386.1 | 4 - 5 | 385 | 104 | |
| GenBank mRNA | AY157834.1 | - | 4 - 5 | 378 | 104 | |
| AY157835.1 | - | 4 - 5 | 378 | 104 | ||
| AY362457.1 | - | 3 - 4 | 230 | 104 | ||
| AY916131.1 | - | 3 - 4 | 221 | 104 | ||
| BC024384.1 | - | 4 - 5 | 379 | 4988988 | 104 | |
| D49949.1 | - | 4 - 5 | 385 | 104 | ||
| DQ316604.1 | - | 3 - 4 | 221 | 104 | ||
| U66244.1 | - | 3 - 4 | 221 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il18
Genomic Map