Assay Details
| Gene Symbol: Irak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177973.1 | NP_001171444.1 | 8 - 9 | 1211 | 63 | |
| NM_001177974.1 | NP_001171445.1 | 8 - 9 | 1208 | 63 | ||
| NM_001177975.1 | NP_001171446.1 | 8 - 9 | 1208 | 63 | ||
| NM_001177976.1 | NP_001171447.1 | 8 - 9 | 1208 | 63 | ||
| NM_008363.2 | NP_032389.2 | 8 - 9 | 1208 | 63 | ||
| GenBank mRNA | AB088370.1 | - | 8 - 9 | 1035 | 63 | |
| AF103876.1 | - | 8 - 9 | 1208 | 63 | ||
| AK045958.1 | - | 8 - 9 | 1117 | 63 | ||
| AK052933.1 | - | 6 - 7 | 1078 | 63 | ||
| AK143694.1 | - | 8 - 9 | 1119 | 63 | ||
| AK144879.1 | - | 8 - 9 | 1117 | 63 | ||
| AY184362.1 | - | 8 - 9 | 1208 | 63 | ||
| AY184363.1 | - | 8 - 9 | 1208 | 63 | ||
| AY184364.1 | - | 8 - 9 | 1208 | 63 | ||
| BC004778.1 | - | 8 - 9 | 1203 | 3257579 | 63 | |
| U56773.2 | - | 8 - 9 | 1032 | 63 | ||
More Information
Set Membership: |
|

Irak1
Genomic Map