Assay Details
| Gene Symbol: Il2ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008367.3 | NP_032393.3 | 6 - 7 | 927 | 81 | |
| GenBank mRNA | AK037369.1 | - | 6 - 7 | 913 | 81 | |
| AK088594.1 | - | 6 - 7 | 869 | 81 | ||
| AK171665.1 | - | 6 - 7 | 864 | 81 | ||
| BC114437.1 | - | 6 - 7 | 833 | 40054387 | 81 | |
| K02891.1 | - | 6 - 7 | 810 | 81 | ||
| M54934.1 | - | 6 - 7 | 730 | 81 | ||
More Information
Set Membership: |
|

Il2ra
Genomic Map