Assay Details
| Gene Symbol: Il2rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008368.4 | NP_032394.1 | 1 - 2 | 119 | 74 | |
| GenBank mRNA | AK017288.1 | - | 1 - 2 | 119 | 74 | |
| AK035289.1 | - | 1 - 2 | 72 | 74 | ||
| AK157572.1 | - | 1 - 2 | 120 | 74 | ||
| AK169481.1 | - | 1 - 2 | 100 | 74 | ||
| BC026869.1 | - | 1 - 2 | 92 | 4015306 | 74 | |
| M28052.1 | - | 1 - 2 | 76 | 74 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK017288.1,AK035289.1,AK157572.1,AK169481.1,BC026869.1,M28052.1,NM_008368.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Citations (1)
Effector and memory CD8+ T cell fate coupled by T-bet and eomesodermin.
- Authors
- Intlekofer AM, Takemoto N, Wherry EJ, Longworth SA, Northrup JT, Palanivel VR, Mullen AC, Gasink CR, Kaech SM, Miller JD, Gapin L, Ryan K, Russ AP, Lindsten T, Orange JS, Goldrath AW, Ahmed R, Reiner SL
- Journal
- Nat Immunol - Page No 1236 (Thu Dec 01 00:00:00 EST 2005)
- Volume (Year)
- 6 (2005)
- Product Usage
- -

Il2rb
Genomic Map