Assay Details
| Gene Symbol: Il2rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008368.4 | NP_032394.1 | 4 - 5 | 437 | 75 | |
| GenBank mRNA | AK017288.1 | - | 4 - 5 | 437 | 75 | |
| AK035289.1 | - | 4 - 5 | 390 | 75 | ||
| AK157572.1 | - | 4 - 5 | 438 | 75 | ||
| BC026869.1 | - | 4 - 5 | 410 | 4015306 | 75 | |
| BC099616.1 | - | 4 - 5 | 411 | 30024496 | 75 | |
| M28052.1 | - | 4 - 5 | 394 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il2rb
Genomic Map