Assay Details
| Gene Symbol: Il2rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008368.4 | NP_032394.1 | 9 - 10 | 1056 | 143 | |
| GenBank mRNA | AK017288.1 | - | 9 - 10 | 1056 | 143 | |
| AK035289.1 | - | 9 - 10 | 1009 | 143 | ||
| AK157572.1 | - | 9 - 10 | 1057 | 143 | ||
| AK169481.1 | - | 9 - 10 | 1037 | 143 | ||
| BC026869.1 | - | 9 - 10 | 1029 | 4015306 | 143 | |
| M28052.1 | - | 9 - 10 | 1013 | 143 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P286496 | BC026869.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P286496 | BQ033566.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P286496 | AK169481.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P286496 | AI047841.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P286496 | AK017288.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P286496 | BC026869.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P286496 | BQ033566.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P286496 | AK169481.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P286496 | AI047841.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P286496 | AK017288.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 14141 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 939607 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 840825 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 645223 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 535030 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 173574 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 432013 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 681071 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 331962 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 750106 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 58348 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 172229 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P286496 | BC026869.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P286496 | BQ033566.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P286496 | AK169481.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P286496 | AI047841.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P286496 | AK017288.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P286496 | BC026869.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P286496 | BQ033566.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P286496 | AK169481.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P286496 | AI047841.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P286496 | AK017288.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P286496 | BC026869.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P286496 | BQ033566.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P286496 | AK169481.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P286496 | AI047841.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P286496 | AK017288.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P286496 | BC026869.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P286496 | BQ033566.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P286496 | AK169481.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P286496 | AI047841.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P286496 | AK017288.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30360417 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30360415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30360425 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30360428 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30360430 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30360435 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30360431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30360421 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30360436 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30360427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30360419 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30360423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30360424 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30360439 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30360433 | - | - |
| Whole Mouse Genome G4122A | A_51_P286496 | BC026869.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P286496 | BQ033566.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P286496 | AK169481.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P286496 | AI047841.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P286496 | AK017288.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P286496 | BC026869.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P286496 | BQ033566.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P286496 | AK169481.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P286496 | AI047841.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P286496 | AK017288.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115737 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12298072 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11989848 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11946941 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11989845 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12298074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30360419 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12299334 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12299337 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12345709 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30360423 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30360428 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12286357 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30360439 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30360413 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12299329 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30360427 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30360452 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12153699 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30360444 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30360447 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30360435 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12211300 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12284289 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30360455 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30360445 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30360465 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30360419 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12209907 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30360415 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30360424 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30360450 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12153711 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30360448 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12299336 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30360433 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30360462 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30360451 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30360457 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30360421 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12211306 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30360461 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10430353 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430349 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430347 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430356 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430354 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430357 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430355 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430345 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430350 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430352 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430348 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430346 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430351 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417546_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448759_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10430344 | - | - |
| Mouse Genome 430 2.0 Array | 1417546_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448759_at | - | - |
| Mouse Genome 430 2.0 Array | 1459870_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1417546_at | - | - |
| Mouse Genome 430 2.0 Array | 1459870_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1448759_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16185_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16185_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500014338.mm.1 | NM_008368.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014338.mm.1 | NM_008368.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014338.mm.1 | TC1500001833.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500014340.mm.1 | NM_008368.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014340.mm.1 | NM_008368.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014340.mm.1 | TC1500001833.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417546_at | - | - |
| Mouse Expression Set 430 | 1448759_at | - | - |
| Mouse Expression Set 430 | 1417546_PM_at | - | - |
| Mouse Expression Set 430 | 1448759_PM_at | - | - |
| Murine Genome U74v2 Set | 100764_at | - | - |
| Murine Genome U74v2 Set | 100764_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417546_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448759_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448759_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417546_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448759_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417546_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417546_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448759_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4730072 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4730072 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004730072 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1246541 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1246541 | - | - |
| mouse-6 v1.1 (Illumina) | scl46990.11.1_92-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4730072 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl46990.11.1_92-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4730072 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1246541 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1246541 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl46990.11.1_92-S | - | - |

Il2rb
Genomic Map