Assay Details
| Gene Symbol: Il9r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134458.1 | NP_001127930.1 | 4 - 5 | 487 | 60 | |
| NM_008374.2 | NP_032400.1 | 4 - 5 | 487 | 60 | ||
| GenBank mRNA | AK088694.1 | - | 4 - 5 | 444 | 60 | |
| BC111909.1 | - | 4 - 5 | 445 | 40046258 | 60 | |
| M84746.1 | - | 4 - 5 | 430 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il9r
Genomic Map