Assay Details
| Gene Symbol: Itgae | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008399.2 | NP_032425.2 | 8 - 9 | 872 | 87 | |
| NM_172944.2 | NP_766532.1 | 8 - 9 | 872 | 87 | ||
| GenBank mRNA | AK040983.1 | - | 8 - 9 | 873 | 87 | |
| AK154021.1 | - | 8 - 9 | 870 | 87 | ||
| BC150690.1 | - | 8 - 9 | 872 | 9087601 | 87 | |
| DQ386608.1 | - | 8 - 9 | 872 | 87 | ||
| U12236.1 | - | 8 - 9 | 896 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Itgae
Genomic Map