Assay Details
| Gene Symbol: Zfp239 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001792.1 | NP_001001792.1 | 4 | 1934 | 67 | |
| NM_008616.2 | NP_032642.2 | 3 | 1754 | 67 | ||
| GenBank mRNA | AK078513.1 | - | 3 | 1748 | 67 | |
| AK133328.1 | - | 1593 | 67 | |||
| BC003315.1 | - | 1 | 688 | 3155528 | 67 | |
| BC080674.1 | - | 1 | 1571 | 67 | ||
| M32057.1 | - | 1 | 1461 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P231630 | M32057.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P231630 | M32057.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020148 | M32057.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020148 | BC080674.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020148 | AK078513.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020148 | AK133328.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020148 | NM_001001792.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 416980 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 492855 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 155491 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 335336 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 526369 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 606424 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 1408 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P130930 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020148 | M32057.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020148 | BC080674.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020148 | AK078513.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020148 | AK133328.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020148 | NM_001001792.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P231630 | M32057.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P231630 | M32057.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P231630 | M32057.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P231630 | M32057.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24783728 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24783723 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24783729 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24783732 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24783727 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24783735 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24783737 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24783738 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24783726 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24783712 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24783736 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24783733 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24783725 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24783721 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24783719 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06041401 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P130930 | - | - |
| Whole Mouse Genome G4122A | A_51_P231630 | M32057.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P231630 | M32057.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132424 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P130930 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05635295 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05635297 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05635296 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05426125 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05426137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24783709 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24783710 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24783733 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24783722 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05458281 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05426124 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24783725 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05426130 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05073976 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24783709 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24783721 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24783764 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24783750 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24783758 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24783746 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05426119 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05837362 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24783767 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24783703 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24783719 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24783766 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24783762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05426137 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24783723 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24783749 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24783748 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24783712 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05382095 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05382096 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24783706 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24783753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24783702 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05570735 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24783738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05382097 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24783744 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24783704 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05426133 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24783711 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24783755 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24783730 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10541103 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10541099 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10541102 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10541100 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10541101 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416586_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10541098 | - | - |
| Mouse Genome 430 2.0 Array | 1416586_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1416586_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22685_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22685_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600010449.mm.1 | TC0600001320.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600010449.mm.1 | NM_001001792.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600010449.mm.1 | NM_001001792.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600010449.mm.1 | NM_008616.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600010449.mm.1 | NM_008616.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600010439.mm.1 | TC0600001320.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600010439.mm.1 | NM_001001792.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600010439.mm.1 | NM_001001792.1 | Refseq |
| Mouse Expression Set 430 | 1416586_PM_at | - | - |
| Mouse Expression Set 430 | 1416586_at | - | - |
| Murine Genome U74v2 Set | 102411_at | - | - |
| Murine Genome U74v2 Set | 102411_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416586_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416586_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416586_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416586_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 464679 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2650215 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 101090725 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002650215 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2436530 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2467917 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2436530 | - | - |
| mouse-6 v1.1 (Illumina) | scl29583.7_500-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0001247.1_46-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101090725 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2650215 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl29583.7_500-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101090725 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2650215 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3127381 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2436530 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3127381 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2467917 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2436530 | - | - |

Zfp239
Genomic Map