Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300023797.mm.1 |
NM_008642.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300023797.mm.1 |
NM_008642.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300023797.mm.1 |
TC0300003016.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300023796.mm.1 |
NM_001163457.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300023796.mm.1 |
NM_001163457.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300023796.mm.1 |
NM_008642.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300023796.mm.1 |
NM_008642.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300023796.mm.1 |
TC0300003016.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300023798.mm.1 |
NM_008642.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300023798.mm.1 |
NM_008642.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300023798.mm.1 |
TC0300003016.mm.1 |
MTA TC |