Assay Details
Gene Symbol: Npr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008727.5 | NP_032753.5 | 2 - 3 | 1275 | 103 | |
GenBank mRNA | BC110659.1 | - | 2 - 3 | 1155 | 30847317 | 103 |
BC139767.1 | - | 2 - 3 | 1134 | 103 | ||
L31932.1 | - | 2 - 3 | 923 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
