Assay Details
| Gene Symbol: Npr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008727.5 | NP_032753.5 | 3 - 4 | 1384 | 95 | |
| GenBank mRNA | BC110659.1 | - | 3 - 4 | 1264 | 30847317 | 95 |
| BC139767.1 | - | 3 - 4 | 1243 | 95 | ||
| J05504.1 | - | 4 - 5 | 1368 | 95 | ||
| L31932.1 | - | 3 - 4 | 1032 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Npr1
Genomic Map