Assay Details
| Gene Symbol: Npr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008728.2 | NP_032754.2 | 6 - 7 | 1449 | 75 | |
| GenBank mRNA | AA763613.1 | - | 1 - 2 | 129 | 75 | |
| D78175.1 | - | 6 - 7 | 1414 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Npr3
Genomic Map