Assay Details
| Gene Symbol: Npy2r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | D86238.1 | - | 1 - 2 | 262 | 96 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the D86238.1 transcripts.
Set Membership: |
|

Npy2r
Genomic Map