Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500018326.mm.1 |
NM_001146161.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018326.mm.1 |
NM_001146161.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018326.mm.1 |
NM_008732.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018326.mm.1 |
NM_008732.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018326.mm.1 |
TC1500002238.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500018327.mm.1 |
NM_001146161.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018327.mm.1 |
NM_001146161.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018327.mm.1 |
NM_008732.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018327.mm.1 |
NM_008732.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018327.mm.1 |
TC1500002238.mm.1 |
MTA TC |