Assay Details
| Gene Symbol: Pax5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008782.2 | NP_032808.1 | 8 - 9 | 1041 | 59 | |
| GenBank mRNA | M97013.1 | - | 8 - 9 | 1041 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pax5
Genomic Map