Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100024423.mm.1 |
NM_001162415.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024423.mm.1 |
NM_001162415.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024423.mm.1 |
NM_001162416.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024423.mm.1 |
NM_001162416.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024423.mm.1 |
NM_008825.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024423.mm.1 |
NM_008825.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024423.mm.1 |
NR_027859.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024423.mm.1 |
NR_027859.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024423.mm.1 |
TC0100003042.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100024428.mm.1 |
NM_008825.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024428.mm.1 |
NM_008825.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024428.mm.1 |
NR_027859.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024428.mm.1 |
NR_027859.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024428.mm.1 |
TC0100003042.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100024429.mm.1 |
NM_008825.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024429.mm.1 |
NM_008825.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024429.mm.1 |
NR_027859.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024429.mm.1 |
NR_027859.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024429.mm.1 |
TC0100003042.mm.1 |
MTA TC |