Assay Details
Gene Symbol: Prkch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008856.3 | NP_032882.2 | 4 - 5 | 737 | 75 | |
GenBank mRNA | AK076352.1 | - | 4 - 5 | 936 | 75 | |
AK145817.1 | - | 4 - 5 | 983 | 75 | ||
AK164044.1 | - | 4 - 5 | 936 | 75 | ||
BC031121.1 | - | 4 - 5 | 737 | 5059633 | 75 | |
D90242.1 | - | 4 - 5 | 674 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
