| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0300001139.mm.1 |
NM_008857.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001139.mm.1 |
NM_008857.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001139.mm.1 |
TC0300000206.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300001138.mm.1 |
NM_008857.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001138.mm.1 |
NM_008857.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001138.mm.1 |
TC0300000206.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300001140.mm.1 |
NM_008857.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001140.mm.1 |
NM_008857.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001140.mm.1 |
TC0300000206.mm.1 |
MTA TC |