Assay Details
Gene Symbol: Ppy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008918.1 | NP_032944.1 | 1 - 2 | 35 | 58 | |
GenBank mRNA | AK131852.1 | - | 1 - 2 | 55 | 58 | |
AK136736.1 | - | 1 - 2 | 55 | 58 | ||
BC099881.1 | - | 1 - 2 | 30 | 40040669 | 58 | |
BC100676.1 | - | 1 - 2 | 30 | 40040664 | 58 | |
BC101946.1 | - | 1 - 2 | 30 | 40040662 | 58 | |
M18208.1 | - | 1 - 2 | 35 | 58 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK131852.1,AK136736.1,BC099881.1,BC100676.1,BC101946.1,M18208.1,NM_008918.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
