Assay Details
Gene Symbol: Prkg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008926.4 | NP_032952.3 | 4 - 5 | 909 | 79 | |
GenBank mRNA | AK038739.1 | - | 4 - 5 | 893 | 79 | |
AK081422.1 | - | 4 - 5 | 910 | 79 | ||
BC113205.1 | - | 4 - 5 | 820 | 40047751 | 79 | |
L12460.1 | - | 4 - 5 | 1181 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
