Assay Details
Gene Symbol: Pspn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008954.2 | NP_032980.2 | 1 - 2 | 159 | 73 | |
GenBank mRNA | AF040960.1 | - | 1 - 2 | 156 | 73 | |
BC130229.1 | - | 1 - 2 | 160 | 40141900 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P312825 | AF040960.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P312825 | BC130229.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P312825 | NM_008954.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P312825 | AF040960.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P312825 | BC130229.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P312825 | NM_008954.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P312825 | AF040960.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P312825 | BC130229.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P312825 | NM_008954.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P312825 | AF040960.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P312825 | BC130229.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P312825 | NM_008954.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P312825 | AF040960.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P312825 | BC130229.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P312825 | NM_008954.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P312825 | AF040960.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P312825 | BC130229.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P312825 | NM_008954.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259492 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259504 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259490 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259497 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259502 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259509 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259507 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259499 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259505 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259493 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259500 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259494 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259496 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259491 | - | - |
Whole Mouse Genome G4122A | A_51_P312825 | AF040960.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P312825 | BC130229.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P312825 | NM_008954.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P312825 | AF040960.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P312825 | BC130229.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P312825 | NM_008954.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122460 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13396938 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13396931 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13396934 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13396930 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13396929 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259451 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259475 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259534 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259502 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259454 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259494 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259540 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259499 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259423 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259509 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259435 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259538 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259466 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259424 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259463 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259542 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259443 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259541 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259493 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259430 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259546 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259492 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259470 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259535 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259438 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259490 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259478 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259545 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259505 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259464 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259442 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259468 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259427 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259445 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259497 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259543 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259496 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259504 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259532 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259473 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259507 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259446 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259472 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259501 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259440 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259550 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259421 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259477 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31259501 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31259509 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13396938 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31259502 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31259493 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31259491 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13396936 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13405705 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31259505 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13228025 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31259499 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31259490 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31259492 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13396934 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10452212 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10452211 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1450375_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10452210 | - | - |
Mouse Genome 430 2.0 Array | 1450375_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1450375_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19197_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19197_at | - | - |
Mouse Expression Set 430 | 1450375_PM_at | - | - |
Mouse Expression Set 430 | 1450375_at | - | - |
Murine Genome U74v2 Set | 100298_at | - | - |
Murine Genome U74v2 Set | 100298_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1450375_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1450375_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450375_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450375_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 577262 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 770520 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 770520 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000770520 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2773560 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2773560 | - | - |
mouse-6 v1.1 (Illumina) | scl49753.2.1_144-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 770520 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl49753.2.1_144-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 770520 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2825886 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2825886 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2773560 | - | - |
