Assay Details
| Gene Symbol: Ptprn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008985.2 | NP_033011.2 | 9 - 10 | 1520 | 76 | |
| GenBank mRNA | AF332083.1 | - | 6 - 7 | 1124 | 76 | |
| AF332084.1 | - | 6 - 7 | 1115 | 76 | ||
| AK041296.1 | - | 9 - 10 | 1526 | 76 | ||
| AK076223.1 | - | 9 - 10 | 1501 | 76 | ||
| AK139398.1 | - | 8 - 9 | 2040 | 76 | ||
| AK220467.1 | - | 9 - 10 | 1478 | 76 | ||
| BC068165.1 | - | 9 - 10 | 1452 | 76 | ||
| DQ832283.1 | - | 9 - 10 | 1497 | 76 | ||
| U11812.1 | - | 9 - 10 | 1438 | 76 | ||
| X74438.1 | - | 9 - 10 | 1537 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptprn
Genomic Map